missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAB1 Antibody (GPN1/2350), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75782-20ug
This item is not returnable.
View return policy
Description
XAB1 Monoclonal antibody specifically detects XAB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| XAB1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/ml, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin : 1-2 μg/ml, Protein Array | |
| Q9HCN4 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| GPN1/2350 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| ATP(GTP)-binding protein, ATPBD1A, GPN-loop GTPase 1, GTPase, MBD2-interacting protein, MBDin, MBDINXPA binding protein 1, XAB1FLJ51176, XPA-binding protein 1 | |
| Recombinant full-length human XAB1 protein (Uniprot: Q9HN4) | |
| 20 μg | |
| Signal Transduction | |
| 11321 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction