missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat C3a ELISA Kit

Product Code. 30245234
Click to view available options
:
96 Tests
This item is not returnable. View return policy

Product Code. 30245234

Brand: Invitrogen™ EEL224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01026
Assay Range 0.63 to 40 ng/mL
Assay Sensitivity 0.38 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Rat
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 24232
Gene Symbol C3
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Complement C3a
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.