missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Granzyme B Quantikine ELISA Kit

Product Code. 30000827
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 30000827

Brand: R&D Systems™ DGZB00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Granzyme B Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Granzyme B Quantikine ELISA Kit quantifies human Granzyme B in cell culture supernates (50 μL).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.30 to 2000 pg/mL
Assay Sensitivity 4.8 pg/mL
Sample Type Cell Culture Supernatant
Specificity Natural and Recombinant Human Granzyme B
For Use With (Application) ELISA
Gene ID (Entrez) 3002
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT
Target Granzyme B
Total Assay Time 3.5 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.