missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Safety-Labeled, Two Color Water Wide-Mouth Wash Bottles
Shop All SP Scienceware Products
Click to view available options
Capacity (English):
16 oz.
Capacity (Metric):
500 mL
Color:
Natural
Autoclavable:
Not Autoclavable
Unit Size:
Pack of 6
Description
Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.
- Easy-to-read printed bottles
- Wide mouth 53 mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 mm
- Toluene is red LDPE bottle
- Polypropylene closure with leakproof design
- Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol
Compliance
OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Natural |
| For Use With (Application) | Distilled Water |
| Material | LDPE, Polyethylene |
| Sterility | Non-sterile |
| Closure Color | Blue |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction