Learn More
Copan Italia X500 Pasteur pipettes 1 ml, graduated 0.25 ml division, sterile, individually packaged, 150 mm long,
Brand:  Copan Italia CP201CS01
X500 Pasteur pipettes 1 ml, graduated 0.25 ml division, sterile, individually packaged, 150 mm long,
Product Code. 18831235
  • £92.98 / Pack of 500