Learn More
Waters X500 Cartridge Oasis with Gilson adapter HLB 30µm30mg, 1mL (pack of 500)
Brand:  Waters WAT058882T4
Encompass
X500 Cartridge Oasis with Gilson adapter HLB 30µm30mg, 1mL (pack of 500)
Product Code. 13082643
  • £1861.50 / Pack of 500
Only null left
Estimated Shipment: 19-01-2026