Learn More
Bohlender™ BOLA™ Spare Knurled Nut
Click to view available options
Description:
Taper 1:10
Taper 1:10/1:10/1:10/1:5
Taper 1:10/1:10/1:5
Type:
Thread M 14
Thread M 7
Thread M 8
Closure Size:
NS 10
NS 12.5, NS 14.5, 13.5
NS 21.5, NS 24.0, NS 29.2, 28.0
Unit Size:
Pack of 20
Pack of 50
Description
- For BOLA™ Stopcocks
- Made of polypropylene
Specifications
Specifications
| Type | Thread M 7 |
| Description | Taper 1:10 |
| Material | Polypropylene |
| Closure Size | NS 10 |
| Color | Red |