Learn More
Agilent Technologies X50 QuEChERS dSPE 15 mL Pigment Sample (EN like) 5
Brand:  Agilent Technologies 5610-2072
Encompass_Preferred
X50 QuEChERS dSPE 15 mL Pigment Sample (EN like) 50 /Pk 885 mg MgSO4 150 mg PSA 15 mg Carbon
Product Code. 30183273
  • £147.15 / Pack of 50
Only null left
Estimated Shipment: 31-12-2025