Learn More
Agilent Technologies X50 Bond Elut C18 cartridge 50 mg 3 mL 40 um 50/pk
Brand:  Agilent Technologies 12105027
Encompass_Preferred
X50 Bond Elut C18 cartridge 50 mg 3 mL 40 um 50/pk
Product Code. 30187016
  • £149.85 / Pack of 50
Only null left
Estimated Shipment: 07-01-2026