missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL4/MIP-1 beta DuoSet ELISA

Product Code. 30000872
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000872 1 Kit (5 Plates) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000872 Supplier R&D Systems™ Supplier No. DY45105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse CCL4/MIP-1 beta DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL4/MIP-1 beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL4/MIP-1 beta.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 - 1000 pg/mL
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 6351
Gene Symbol Ccl4
Host Species Mouse
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target CCL4/MIP-1 beta
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.