missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL3/MIP-1 alpha DuoSet ELISA
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
Description
The Mouse CCL3/MIP-1 alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL3/MIP-1 alpha.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 7.81 to 500 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 5 96-well Plates |
| For Use With (Application) | ELISA |
| Synonym | C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction