missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical X4 Kit of ethanol reference solutions for alcoholcontent measuring instruments.
BIOPHARMA

Brand:  Fisher Chemical KES 1A


X4 Kit of ethanol reference solutions for alcoholcontent measuring instruments.

Product Code. 15769819

  • ONLINE EXCLUSIVE PRICE
    £262.00
    £257.00 / Pack of 4
    Save £5.00
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Kit of ethanol reference solutions for alcohol content measuring instruments; 5%, 9%, 11.5%,

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 1
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.