Learn More
Epredia X4 FORMALDEHYDE, BUFFERED,SOLUTION, 10%, STABILISE
Brand: Epredia 9400-1
X4 FORMALDEHYDE, BUFFERED,SOLUTION, 10%, STABILISEd, neutral buffered 3.79L
Description
Safety and Handling
- 10% Neutral Buffered Formalin
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, methanol, sodium phosphatedibasic, sodium acid phosphate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.