Learn More
Agilent Technologies X3 ZORBAX RRHD Eclipse Plus Phe nyl-Hexyl 3.0 x 10
Brand:  Agilent Technologies 959758-312K
Encompass_Preferred
X3 ZORBAX RRHD Eclipse Plus Phe nyl-Hexyl 3.0 x 100 mm 1.8 um 1200 bar M VK 3/pk
Product Code. 30189162
  • £2786.40 / Pack of 3
Only null left
Estimated Shipment: 06-01-2026