Learn More
Invitrogen™ Gata-3 Monoclonal Antibody (TWAJ), Brilliant Violet™ 605, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand:  Invitrogen™ 406-9966-41
 View more versions of this product
Product Code. 30019897
  • £158.00 / Pack of 25
Description
Gata-3 Monoclonal antibody specifically detects Gata-3 in Human, Mouse, Porcine, Rhesus Monkey samples. It is validated for Flow Cytometry
The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gata-3
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P23771, P23772
GATA3
Affinity chromatography
RUO
14462, 2625, 713840, 733631
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
TWAJ
Brilliant Violet 605
GATA3
EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3
Rat
25 Tests
Primary
Human, Mouse, Pig, Rhesus Macaque
Antibody
IgG2b κ