missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments X25 Buffer Hanna Technical pH 1.0 20mL sachet
Brand: Hanna Instruments HI50001-02
X25 Buffer Hanna Technical pH 1.0 20mL sachet
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ph 1.00 Buffer Solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction