missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Prepared Sabouraud Dextrose Broth Bottled Media

Product Code. 10045972
Change view
Click to view available options
Fill Weight:
10.0 to 10.3 g
100-101.5g
9.5 to 10.5 g
Format:
100 mL DIN bottle
Tube
Universal Bottle
Quantity:
10/Pk.
24/Pk.
50/Pk.
Unit Size:
Each
Pack of 24
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format Quantity unitSize
10045972 10.0 to 10.3 g Universal Bottle 24/Pk. Pack of 24
12766899 100-101.5g 100 mL DIN bottle 10/Pk. Each
13225809 9.5 to 10.5 g Tube 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10045972 Supplier Thermo Scientific™ Supplier No. BO0358E

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Confidently perform sterility testing and/or determining the fungistatic activity of pharmaceutical products with this broth.

A non-selective broth for confidently performing sterility testing and/or determining the fungistatic activity of pharmaceutical products.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 25°C
Format Universal Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 10.0 to 10.3 g
Packaging Type 24 Universal Bottles with Screw Cap Closures/Box
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.