missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ 360 Isolator Cleaning System
BIOPHARMA

Product Code. 17543286 Shop All Berkshire Products
Change view
Click to view available options
Product Type:
Isolater Cleaning Tool
Isolator Cleaning Tool Cover
Material:
Knitted Polyester Elastic
Polyester Nonwoven Elastic, Cellulose
Stainless Steel, Aluminum
Quantity:
1 Ea.
10/Pk.
6/Pk.
Unit Size:
Each
Pack of 200
Pack of 25
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Material Quantity unitSize
17543286 Isolator Cleaning Tool Cover Knitted Polyester Elastic 10/Pk. Pack of 200
15428500 Isolater Cleaning Tool Stainless Steel, Aluminum 1 Ea. Each
15418500 Isolator Cleaning Tool Cover Knitted Polyester Elastic 10/Pk. Pack of 5
17503286 Isolator Cleaning Tool Cover Knitted Polyester Elastic 6/Pk. Pack of 25
15438500 Isolator Cleaning Tool Cover Polyester Nonwoven Elastic, Cellulose 10/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17543286 Supplier Berkshire™ Supplier No. EC360.ECST.20
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Quick, easy and efficient cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.

EasyClean 360 ICT Sterile Knitted Polyester Elastic and Padded Covers (17-987-362)

  • Made from Berkshire's SuperPolx™ 1200A 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior giver, particle and residue performance
  • Non-shedding and low-linting cover per USP requirements
  • Stretch fabric and elastic edge for easy on/off with secure fit on mop head
  • Absorbent polyester knit is abrasion resistant; handles a wide range of cleaning solutions
  • Compatible for use in ISO Class 3/EU Grade A Environments
  • Sterile

Specifications

Product Type Isolator Cleaning Tool Cover
Color White
Sterility Sterile
Autoclavable Autoclavable
Quantity 10/Pk.
Includes 10 sterile knitted polyester elastic covers and 1 sterile knitted polyester padded cover/pack; 20 packs/case
For Use With (Equipment) EasyClean™ 360 Isolator Cleaning Tool
Description Mop Covers
For Use With (Application) Cleaning of isolators, biological safety cabinets, laminar flow hoods and other mini environments.
Certifications/Compliance ISO Class 3/EU Grade A
Material Knitted Polyester Elastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.