missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon, Low Alloy Steel Set, 20 pieces, Ø38x3mm, ARMI∣MBH
Click to view available options
Unit Size:
Pack of 20
Description
Carbon, Low Alloy Steel Set, 20 pieces, Ø38x3mm
Specifications
Specifications
| CAS | 7439-89-6 |
| Form | Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction