missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Tangential Flow Filtration (TFF)-Large EV Isolation Adaptor
Shop All Bio Techne Products
Click to view available options
:
2 Units
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Quantity | 2 Units |
| Sample Type | EV containing matrices (urine, cell culture supernatant, plasma, serum, plant extracts) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction