missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat FGF-acidic (FGF-1) Recombinant Protein, PeproTech®
Click to view available options
Quantity:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Pack of 2
Description
400-29A-1MG will be provided as 2 x 500 μg (400-29A-500UG). Recombinant Rat FGF-acidic is a 15.9 kDa protein consisting of 141 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
Specifications
Specifications
| Accession Number | P61149 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 25317 |
| Molecular Weight (g/mol) | 15.9 kDa |
| Name | Rat FGF-acidic (FGF-1) |
| Quantity | 2 x 500 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction