missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formoterol Fumarate Dihydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Formoterol Fumarate Dihydrate
Specifications
Specifications
| Chemical Name or Material | Formoterol fumarate dihydrate |
| CAS | 183814-30-4 |
| Analyte or Component Names | Formoterol fumarate dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2[C19 H24 N2 O4] . C4 H4 O4 . 2[H2O] |
| Certifications/Compliance | ISO 9001 |
| Synonym | Formoterol Fumarate Dihydrate, Formamide, N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-, rel-, (2E)-2-butenedioate (2:1) (salt), dihydrate (9CI), Formamide, N-[2-hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-, (R*,R*)-, (E)-2-butenedioate (2:1) (salt), dihydrate |
| InChI Formula | InChI=1S/2C19H24N2O4.C4H4O4.2H2O/c2*1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-3(6)1-2-4(7)8;;/h2*3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2H,(H,5,6)(H,7,8);2*1H2/b;;2-1+;;/t2*13-,19+;;;/m11.../s1 |
| SMILES | O.COc1ccc(C[C@@H](C)NC[C@H](O)c2ccc(O)c(NC=O)c2)cc1.COc3ccc(C[C@@H](C)NC[C@H](O)c4ccc(O)c(NC=O)c4)cc3.OC(=O)\C=C\C(=O)O |
| IUPAC Name | fumaric acid;N-[2-hydroxy-5-[rac-(1R)-1-hydroxy-2-[[rac-(1R)-2-(4-methoxyphenyl)-1-methyl-ethyl]amino]ethyl]phenyl]formamide;hydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion