Learn More
Sarstedt X1000 Tube with cap 8ml, 57x16.5mmFlat Base, PP with assembled cap, sterile
Brand:  Sarstedt 60.542.024
Encompass
X1000 Tube with cap 8ml, 57x16.5mmFlat Base, PP with assembled cap, sterile
Product Code. 15574169
  • £294.87 / Pack of 1000
Only null left
Estimated Shipment: 22-04-2026