Learn More
Thermo Scientific Menzel X1000 Coverslip 24x60mm #1.5 (0.16-0.19mm)
Brand: Thermo Scientific Menzel BB02400600AC13MNZ0
66.04 GBP valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24145" to get your promotional price.
X1000 Coverslip 24x60mm #1.5 (0.16-0.19mm)