Learn More
Gerstel X100 Screw Cap, 9mm, Silicone blue /PTFE white, 45° Sho re A, 1.3mm, 100/pk
Brand:  Gerstel 093640-072-00
Encompass_Preferred
X100 Screw Cap, 9mm, Silicone blue /PTFE white, 45° Sho re A, 1.3mm, 100/pk
Product Code. 30224190
  • £85.19 / Pack of 100
Only null left
Estimated Shipment: 08-01-2026