missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse MIP-2 (CXCL2) ELISA Development Kit (ABTS), PeproTech®

Product Code. 30089202
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 30089202

Brand: Invitrogen™ 900K152K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10889
Assay Range Lot-specific
Assay Sensitivity Lot-specific
Conjugate HRP
Product Type ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20310
Gene Symbol Cxcl2
Kit Contents Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Storage Requirements Refer to product documentation for component specific storage temperature.
Target CXCL2
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.