missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MCP-2/CCL8 ELISA Development Kit (ABTS), PeproTech®
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications
Specifications
| Accession Number | P80075 |
| Assay Range | Lot-specific |
| Assay Sensitivity | Lot-specific |
| Conjugate | HRP |
| Product Type | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6355 |
| Gene Symbol | CCL8 |
| Kit Contents | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Label or Dye | HRP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction