missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EG-VEGF/PROK1 ELISA Development Kit (ABTS), PeproTech®
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
Pack of 10
Description
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
Specifications
Specifications
| Accession Number | P58294 |
| Assay Range | Lot-specific |
| Assay Sensitivity | Lot-specific |
| Conjugate | HRP |
| Product Type | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 84432 |
| Gene Symbol | PROK1 |
| Kit Contents | Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate |
| Label or Dye | HRP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction