Learn More
Agilent Technologies X1 VF624ms,fs,5in 0.25mm,60m,1. 4um,1/pk
Brand:  Agilent Technologies CP9103I5
Encompass_Preferred
X1 VF624ms,fs,5in 0.25mm,60m,1. 4um,1/pk
Product Code. 30189056
  • £1825.20 / Each
Only null left
Estimated Shipment: 07-01-2026