Learn More
Agilent Technologies X1 Rectangular macro cell optic al glass open top
Brand:  Agilent Technologies 6610016400
Encompass_Preferred
X1 Rectangular macro cell optic al glass open topwi th PTFE lid supplied 50 mm pathlength 17
Product Code. 30188784
  • £163.35 / Each
Only null left
Estimated Shipment: 07-01-2026