Learn More
Agilent Technologies X1 Quinine sulfate 1 PPM in wat er 3 x 2 mL ampule
Brand:  Agilent Technologies 5190-0522
Encompass_Preferred
X1 Quinine sulfate 1 PPM in wat er 3 x 2 mL ampules for OQ of LC systems
Product Code. 30189970
  • £101.25 / Each