Learn More
Agilent Technologies X1 PoraPak Q 80/100 mesh 1.8 m x 1/8 inch 2 mm sta
Brand:  Agilent Technologies G3591-70010
Encompass_Preferred
X1 PoraPak Q 80/100 mesh 1.8 m x 1/8 inch 2 mm stai nless steel packed G C column
Product Code. 30193309
  • £309.15 / Pack of 1
Only null left
Estimated Shipment: 14-04-2026