Learn More
Agilent Technologies X1 J&W HP-PLOT Q PT GC Column 3 0 m 0.53 mm 40.00
Brand:  Agilent Technologies 19095P-QO4PT
Encompass_Preferred
X1 J&W HP-PLOT Q PT GC Column 3 0 m 0.53 mm 40.00um with two particle t raps (front and rear
Product Code. 30183550
  • £1246.05 / Each
Only null left
Estimated Shipment: 31-12-2025