Learn More
Agilent Technologies X1 J&W HP-5MS GC column 30 m 0. 25 mm 0.25 um with
Brand:  Agilent Technologies 19091S-433-KEY
Encompass_Preferred
X1 J&W HP-5MS GC column 30 m 0. 25 mm 0.25 um with s mart key for 8890 GC system
Product Code. 30188216
  • £1003.05 / Each
Only null left
Estimated Shipment: 07-01-2026