Learn More
Agilent Technologies X1 J&W DB-624 Ultra Inert GC Co lumn 30 m 0.32 mm1. 80 um 7 inch cage
Brand:  Agilent Technologies 123-1334UI
Encompass_Preferred
X1 J&W DB-624 Ultra Inert GC Co lumn 30 m 0.32 mm1. 80 um 7 inch cage
Product Code. 30185080
  • £1113.75 / Pack of 1
Only null left
Estimated Shipment: 14-04-2026