Learn More
Agilent Technologies X1 J&W CP-Sil 5 CB for Sulfur G C Column 30 m 0.32
Brand:  Agilent Technologies CP7529
Encompass_Preferred
X1 J&W CP-Sil 5 CB for Sulfur G C Column 30 m 0.32 m m 4.00 um 7 inch cag e
Product Code. 30185009
  • £997.65 / Each
Only null left
Estimated Shipment: 07-01-2026