Learn More
Agilent Technologies X1 J&W CP-Select 624 CB GC Colu mn 30 m 0.25 mm 1.
Brand:  Agilent Technologies CP7412
Encompass_Preferred
X1 J&W CP-Select 624 CB GC Colu mn 30 m 0.25 mm 1.40 um 7 inch cage
Product Code. 30184373
  • £957.15 / Each
Only null left
Estimated Shipment: 06-01-2026