Learn More
Agilent Technologies X1 InfinityLab Quick Change val ve head 2-position
Brand:  Agilent Technologies 5067-4283
Encompass_Preferred
X1 InfinityLab Quick Change val ve head 2-position/1 0-port maximum press ure 800 bar
Product Code. 30185219
  • £5583.60 / Each
Only null left
Estimated Shipment: 29-12-2025