Learn More
Agilent Technologies X1 HayeSep Q 80/100 mesh 2 m x 1/8 inch 2 mm stain
Brand:  Agilent Technologies G3591-70005
Encompass_Preferred
X1 HayeSep Q 80/100 mesh 2 m x 1/8 inch 2 mm stainl ess steel packed GC column
Product Code. 30185751
  • £313.20 / Each
Only null left
Estimated Shipment: 07-01-2026