Learn More
Agilent Technologies X1 GC packed column UltiMetal 0 .33 ft (0.1 m) 1/1
Brand:  Agilent Technologies CP915192
Encompass_Preferred
X1 GC packed column UltiMetal 0 .33 ft (0.1 m) 1/16 in OD 1 mm OD Lowox phase on Chromosorb
Product Code. 30185525
  • £1264.95 / Each
Only null left
Estimated Shipment: 08-01-2026