Learn More
Agilent Technologies X1 GC packed column in Nickel t ubing length 9 ft
Brand:  Agilent Technologies G3591-82064
Encompass_Preferred
X1 GC packed column in Nickel t ubing length 9 ft(2 .74 M) 1/8 in. OD 2 mm ID MolSieve 5A pa
Product Code. 30186038
  • £460.35 / Pack of 1
Only null left
Estimated Shipment: 20-04-2026