Learn More
Agilent Technologies X1 CE extended light path (3x) capillary PVA 50 um
Brand:  Agilent Technologies G1600-61240
Encompass_Preferred
X1 CE extended light path (3x) capillary PVA 50 um id 24.5 cm effective length 33 cm total
Product Code. 30189616
  • £527.85 / Each
Only null left
Estimated Shipment: 07-01-2026