Learn More
BRAND™ X-Shape PTFE Magnetic Stirring Bars
An excellent option for liquid agitation in the laboratory.
Brand:  BRAND™ 137634
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10508221
  • £71.50 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Steady spinning position
  • Excellent stirring action
  • Provided in packs of 10
Specifications
Specifications
25 mm
10/Pk.
10 mm
10mm
PTFE
Cross