Learn More
Draeger X-plore™ 1700 Single-use Respirator FFP3 With Valve
Description
- CoolSAFE™ filter material combines low breathing resistance and high filter performance
- Flexible nose clip ensures good fit in critical areas
- Soft inner layer is hypoallergenic, repels moisture for longer periods
- VarioFLEX™ head harness with continuous loop makes it easy to don and doff, soft, tearproof eleastic is easily adjusted and prevents hair entanglement
- Optional CoolMAX™ exhalation valve offers low breathing resistance, prevents heat build-up and helps you stay cool
- Foldflat form saves space and makes them easier to transport
Specifications
Specifications
| Description | FFP3 valved respirator facemask with optional CooplMAX™ exhalation valve |
| Filter Type | FFP3 |
| Material | Hypoallergenic inner material |
| With Exhalation Valve | Yes |
| Filtration Layer Material | High performance CoolSAFE filter material, additional dolomite dust testing requirements achieved |
| Material Headstrap | VarioFLEX continuous loop adjustable elastic head harness |
| Nosepiece Type | Flexible nose clip |