Learn More
Melford X-Phos BCIP
Product Code. 30018114
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018114 100 mg 100mg
30018115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018114 Supplier Melford Supplier No. B740000.1
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques.
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.
Specifications
Quantity 100 mg