Learn More
Abeba™ X-Light Safety Loafer
Product Code. 17651915
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17651915 39 1 pair
17611915 35 1 pair
17621915 36 1 pair
17631915 37 1 pair
17641915 38 1 pair
17661915 40 1 pair
17671915 41 1 pair
17681915 42 1 pair
17691915 43 1 pair
17601925 44 1 pair
17611925 45 1 pair
17621925 46 1 pair
17631925 47 1 pair
17641925 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17651915 Supplier Abeba™ Supplier No. 711037/39
Stock Up Your Lab
Use promo code 28007
View all promotions
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Abeba X-Light Black Safety Loafer - Smooth leather safety shoes with elastic instep.
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Closure Type Elastic Band
Color Black
Material Leather
Size 39
Toe Material Steel
Certifications/Compliance (Europe) CE, EN ISO 20345:2011, S2, SRC