Learn More
ABEBA™ X-Light Black/Yellow Safety Trainers S3 ESD
Black/yellow X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
Brand:  ABEBA™ 7131860/46
Additional Details : Weight : 1.01000kg
 View more versions of this product
Product Code. 17171575
  • £85.86 / 1 pair
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Lightweight, comfortable occupational shoes
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black and yellow Cordura
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Specifications
Black, Yellow
46
Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Trainers
S3 SRC ESD
Cordura
Steel
Unisex
Water Resistant