Learn More
ABEBA™ X-Light Black/Yellow Safety Trainers S3 ESD
Product Code. 17151575
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17151575
Brand: ABEBA™ 7131860/44
Only null left
Add to basket
Add to basket
Black/yellow X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
  • Lightweight, comfortable occupational shoes
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black and yellow Cordura
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color Black, Yellow
Material Cordura
Size 44
Toe Material Steel
Certifications/Compliance (Europe) Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Gender Unisex
Ankle Style Trainers
Waterproof Water Resistant
Product Type S3 SRC ESD