Learn More
ABEBA™ X-Light Black Safety Trainers S3 ESD
Product Code. 17181585
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17181585 43 1 pair
17101585 35 1 pair
17111585 36 1 pair
17101595 45 1 pair
17111595 46 1 pair
17121595 47 1 pair
17131595 48 1 pair
17121585 37 1 pair
17131585 38 1 pair
17141585 39 1 pair
17151585 40 1 pair
17161585 41 1 pair
17171585 42 1 pair
17191585 44 1 pair
14 options
Product Code. 17181585
Brand: ABEBA™ 7131874/43
Only null left
Add to basket
Add to basket
Black X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
  • Lightweight, comfortable safety trainer
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black smooth leather
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color Black
Material Leather
Size 43
Toe Material Steel
Certifications/Compliance (Europe) Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Gender Unisex
Ankle Style Low-cut
Waterproof Water Resistant
Product Type S3 SRC ESD