missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress X-GAL
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
Specifications
Specifications
| CAS | 7240-90-6 |
| Solubility | DMSO : 100 mg/mL (244.72 mM∣Need ultrasonic) |
| Synonym | BCIG |
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Quantity | 200 mg |
| Molecular Weight (g/mol) | 408.63 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 95.66% |
| Shipping Condition | Room temperature |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction